Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
Brand:  Cole-Parmer™ 3053820
Additional Details : Weight : 0.02270kg
 View more versions of this product
Product Code. 16609285
  • 2130.00 SEK / Each
Estimated Shipment: 27-09-2022
to see stock.
Add to basket
Description
Description
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Specifications
Specifications
1.27 cm
PTFE
PTFE Gasket
20