Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
Brand: Cole-Parmer™ 3053868
Additional Details : Weight : 0.04540kg
Description
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Specifications
5.08 cm | |
PTFE | |
PTFE Gasket | |
60 |