Läs mer
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)
Beskrivning
- Air leak rate <10-8 to 10-6 mbar l/s (high vacuum, overpressure).
- Solvent leak rate in containers <0.1 to 0.03mg/d.
- Temperature resistant from -200 to +300°C.
- The elastic ring (0.07mm thick) adheres to the ground joint without a groove, is reusable and fits to ground joint clamps.
- Other sizes on request.
Specifikationer
Specifikationer
| Material | PTFE |
| Storlek | NS 14 |