Brand™ PTFE Stirring Rod
Product Details
Brand: Brand™
Manufacturer Part Number: 136210
UNSPSC: 41120000
Additional Details:
Additional Details: Weight: 0.02000kg
Product Code. 10003950
Quantity Price
1 122.4  SEK / Each
Estimated Shipment
30-05-2017
Add to basket
Description and Specification
Specification
Diameter (Metric) 8mm
Case Quantity 1Each
Packaging 1Pack
Material PTFE
Length (Metric) 150mm