Läs mer
Fisherbrand™ PTFE Stopper, Flask with Handle
Fisherbrand™ PTFE Stopper, Flask with Handle are for use with volumetric flasks and similar containers.
Brand: Fisherbrand™ 010.4.16
Beskrivning
- Solid Construction
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifikationer
Fisherbrand™ PTFE Stopper, Flask with Handle, White Handle | |
1 | |
-200°C to +280°C | |
PTFE |
Standard Taper Ground Glass Joints | |
16 (US) | |
White |
Korrigering av produktinnehåll
Produkttitel
Fisherbrand™ PTFE Stopper, Flask with Handle > Size (US): 16; Handle Color: Blue
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering