Learn More
BRAND™ PTFE Watch Glass
PTFE Watch Glasses are ideally suited for covering beakers.
Brand:  BRAND™ 150910
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 10303771
  • 203.00 SEK / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Constructed of PTFE
  • Ideal for covering PTFE beakers
  • Excellent chemical resistance
Specifications
Specifications
PTFE
Watch Glass
Beaker
75 mm