missing translation for 'onlineSavingsMsg'
Learn More

PTGER4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24895-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16813405

  • 4721.67 SEK / 150µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. Has a relaxing effect on smooth muscle. May play an important role in regulating renal hemodynamics, intestinal epithelial transport, adrenal aldosterone secretion, and uterine function.
Specifications

Specifications

PTGER4
Polyclonal
Unconjugated
PTGER4
EP4, EP4R, PGE receptor EP4 subtype, PGE2 receptor EP4 subtype, Prostanoid EP4 receptor, PTGER2, PTGER4
Rabbit
Antigen Affinity Chromatography
RUO
19219, 5734, 84023
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P32240, P35408, P43114
PTGER4
PTGER4 Fusion Protein Ag19217
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers