missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGER4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24895-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. Has a relaxing effect on smooth muscle. May play an important role in regulating renal hemodynamics, intestinal epithelial transport, adrenal aldosterone secretion, and uterine function.Specifications
PTGER4 | |
Polyclonal | |
Unconjugated | |
PTGER4 | |
EP4, EP4R, PGE receptor EP4 subtype, PGE2 receptor EP4 subtype, Prostanoid EP4 receptor, PTGER2, PTGER4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19219, 5734, 84023 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P32240, P35408, P43114 | |
PTGER4 | |
PTGER4 Fusion Protein Ag19217 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |