Learn More
PTPIP51 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20641-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
RMDN3 is a protein coding gene involved in cellular calcium homeostasis regulation. May participate in differentiation and apoptosis of keratinocytes. Overexpression induces apoptosis.Specifications
PTPIP51 | |
Polyclonal | |
Unconjugated | |
RMDN3 | |
FAM82A2, FAM82C, hRMD 3, Protein FAM82A2, Protein FAM82C, PTPIP51, RMD 3, RMD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55177 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96TC7 | |
RMDN3 | |
PTPIP51 Fusion Protein Ag14708 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |