Learn More
PTPN7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15286-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16824404
  • 4721.67 SEK / 150µL
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This gene is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The noncatalytic N-terminus of this PTP can interact with MAP kinases and suppress the MAP kinase activities. This PTP was shown to be involved in the regulation of T cell antigen receptor signaling, which was thought to function through dephosphorylating the molecules related to MAP kinase pathway. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PTPN7
Polyclonal
Unconjugated
PTPN7
BPTP 4, HEPTP, LC PTP, LPTP, PTPN7, PTPNI
Rabbit
Antigen Affinity Chromatography
RUO
246781, 320139, 5778
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35236, P49445, Q8BUM3
Ptpn7
PTPN7 Fusion Protein Ag7453
150 μL
Primary
Human, Mouse, Rat