missing translation for 'onlineSavingsMsg'
Learn More

PTPRZ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55125-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826585

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to PTPRZ1.

Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important signaling cascades. .
Specifications

Specifications

PTPRZ1
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B9EKR1, P23471, Q62656
HPTPZ, HPTPzeta, HTPZP2, phosphacan, PTP ZETA, PTP18, PTPRZ, PTPRZ1, PTPRZ2, PTPZ, R PTP zeta, R PTP zeta 2, RPTPB, RPTPbeta
Rabbit
IgG
150 μL
-20°C
Primary
19283, 25613, 5803
Western Blot
Unconjugated
PTPRZ1
Liquid
PTPRZ1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers