missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ PTX3 Recombinant Rabbit Monoclonal Antibody (10D9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550008
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PTX3 Recombinant Monoclonal Antibody for Flow, ELISA
Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of 'long pentraxins', which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
Specifikationer
| PTX3 | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26022 | |
| PTX3 | |
| A synthesized peptide derived from human PTX3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry | |
| 10D9 | |
| Unconjugated | |
| PTX3 | |
| AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5806 | |
| -20°C or -80°C if preferred | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering