missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUF60 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10810-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
PUF60 encodes a protein that is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene.Specifications
PUF60 | |
Polyclonal | |
Unconjugated | |
PUF60 | |
FBP interacting repressor, FIR, PUF60, Ro binding protein 1, RoBP1, RoBPI, Siah binding protein 1, Siah BP1, SIAHBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22827, 67959, 84401 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UEB3, Q9UHX1, Q9WV25 | |
Puf60 | |
PUF60 Fusion Protein Ag1053 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |