missing translation for 'onlineSavingsMsg'
Learn More

Pullulan, MP Biomedicals™

Catalog Number 11447392
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
9057-02-7
(C18H30O15)AHO
GFZFEWWPMNSVBS-WVZDODFGSA-N
Pullulan from Aureobasidium pullulans
131636581
(2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol
COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O
This item is not returnable. View return policy
9057-02-7
(C18H30O15)AHO
GFZFEWWPMNSVBS-WVZDODFGSA-N
Pullulan from Aureobasidium pullulans
131636581
(2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol
COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O

Produced from starch by cultivating a black yeast. Pullulan, MP Biomedicals is an edible plastic with excellent adhesiveness and film formability. It is a natural neutral polysaccharfied used in studies assessing bone tissue engineering and investigating

Quantity 5 g
Solubility Information Dissolves easily in cold and hot water (50mg/mL; 1/20: Clear).
Formula Weight 200,000 (Lit.)
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'