missing translation for 'onlineSavingsMsg'
Learn More

PUM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11586-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878763

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
Specifications

Specifications

PUM2
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q80U58, Q8TB72
KIAA0235, PUM2, PUMH2, Pumilio 2, Pumilio homolog 2, pumilio homolog 2 (Drosophila), PUML2
Rabbit
IgG
20 μL
-20°C
Primary
23369, 298874, 80913
Western Blot
Unconjugated
PUM2
Liquid
Pum2
PUM2 Fusion Protein Ag2179
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers