Läs mer
Punicalagin, TRC
Produktkod. 30422806
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30422806 10 mg
1 options
Produktkod. 30422806 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP84027010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Punicalagins A&B
CAS 65995-63-3
Molekylformel C48 H28 O30
Kvantitet 10 mg
Synonym Punicalagin, Cyclic 4,6-[2,2'-(5,10-Dihydro-2,3,7,8-tetrahydroxy-5,10-dioxo[1]benzopyrano[5,4,3-cde][1]benzopyran-1,6-diyl)bis[3,4,5-trihydroxybenzoate]] cyclic 2,3-(4,4',5,5',6,6'-hexahydroxy[1,1'-biphenyl]-2,2'-dicarboxylate)','[2(S),4(S,S)]-
InChI formel InChI=1S/C48H28O30/c49-5-16-40(76-46(70)9-3-13(52)28(56)32(60)18(9)17-8(44(68)74-16)2-12(51)27(55)31(17)59)39-15(54)6-73-43(67)7-1-11(50)29(57)33(61)19(7)21-25-23-24-26(48(72)78-41(23)37(65)35(21)63)22(36(64)38(66)42(24)77-47(25)71)20-10(45(69)75-39)4-14(53)30(58)34(20)62/h1-5,15-16,39-40,50-66H,6H2
LEDER OC1COC(=O)c2cc(O)c(O)c(O)c2c3c(O)c(O)c4OC(=O)c5c(c(O)c(O)c6OC(=O)c3c4c56)c7c(O)c(O)c(O)cc7C(=O)OC1C8OC(=O)c9cc(O)c(O)c(O)c9c%10c(O)c(O)c(O)cc%10C(=O)OC8C=O
Molekylvikt (g/mol) 1084.72
Formel vikt 1084.0665