Learn More
Labcon™ PurePlus™ 1.2 mL Sample Library Tubes
Sample library tubes come in strips of 8, 12, or as individual tubes.
Brand: Labcon™ 3917-800-000-9
Additional Details : Weight : 0.60320kg
Description
- Available in 96-place racks or resealable bags
- ATP, RNase/DNase, Endotoxin-free (non-pyrogenic)
Specifications
Autoclavable | |
Polypropylene | |
80/Pk. | |
12 mL |
Clear | |
Strips of 12, Bagged | |
80 Tubes/Pk., 10 Pk./Cs. | |
Non-sterile |