Learn More
Brand™ Pursept™ A Xpress Surface Disinfecting Detergents
Disinfects in just 15 seconds. BRAND™ Pursept™ A Xpress Surface Disinfecting Detergents are effective against bacteria and fungi with little organic burden with mechanics. Limited virucidal efficacy including HIV, HBV, and HCV after 15 seconds.
1445.00 SEK - 3415.00 SEK
Specifications
Product Type | Disinfectant Detergent |
---|---|
Form | Liquid |
pH | 7.5 |
Appearance | Colorless |
Certifications/Compliance | DGHM/VAH Tested and RKI Compliant |
Product Code | Brand | Volume (Metric) | Container Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Container Type | Price | Quantity & Availability | ||||
10394741
|
Brand™
44835 |
5 L | Can |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11814110
|
Brand™
44830 |
1 L | Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Disinfects in just 15 seconds. Refillable pump-atomizer without gas. Pleasant, fresh fragrance.- DGHM/VAH tested and RKIcompliant, with aerosol-free application (disinfectant wipes).
100 g Pursept™ A Xpress contain: 55 g ethanol, 0.03 g n-alkyl-aminopropyl-glycine, auxiliary agents.
Chemical and physical specifications:
- Clear, colorless liquid,
- Density: approx. 0.9 g/ml, pH value: approx. 7.5.
- Surface disinfection: Bacteria and fungi with little organic burden with mechanics acc. to DGHM/VAH directives: 15 seconds.
- Limited virucidal efficacy* (incl. HIV, HBV, HCV): 15 seconds.
- Effectiveness against TbB: 30 seconds.
Specifications
Disinfectant Detergent | |
7.5 | |
DGHM/VAH Tested and RKI Compliant | |
With Aerosol-Free Application |
Liquid | |
Colorless | |
100g Pursept™ A Xpress Contain: 55g Ethanol, 0.03g n-Alkyl-Aminopropyl-Glycine, Auxiliary Agen |