Learn More
Ark-Plas Products™ PVC Hose
Made of polyvinyl chloride. Ark-Plas Products™ PVC Hose is ideal for use with connecting hoses of various diameters. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies.
Brand:  Ark-Plas Products™ TCX22-FC5
Additional Details : Weight : 0.23000kg
 View more versions of this product
Product Code. 11737689
  • 393.53 SEK / Each
Estimated Shipment
25-05-2021
Add to basket
Description
Description
  • Chemical and corrosion resistance
  • Abrasion and wear resistance
  • Rubber-like flexibility
Specifications
Specifications
Green
PVC
1/32 in.
1/16 in.
1/8 in.