Learn More
Ark-Plas Products™ PVC Hose
Made of polyvinyl chloride. Ark-Plas Products™ PVC Hose is ideal for use with connecting hoses of various diameters. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies.
Brand:  Ark-Plas Products™ TCX27-FC1
Additional Details : Weight : 0.82800kg
Product Code. 11757539
  • 1435.00 SEK / Each
In Stock
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
  • Chemical and corrosion resistance
  • Abrasion and wear resistance
  • Rubber-like flexibility
Specifications
Specifications
1/8 in.
1/4 in.
Black
PVC
1/16 in.