Learn More
Ark-Plas Products™ PVC Hose
Made of polyvinyl chloride. Ark-Plas Products™ PVC Hose is ideal for use with connecting hoses of various diameters. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies.
Brand: Ark-Plas Products™ TCX27-FC1
Additional Details : Weight : 0.82800kg
Description
- Chemical and corrosion resistance
- Abrasion and wear resistance
- Rubber-like flexibility
Specifications
1/8 in. | |
1/4 in. | |
Black |
PVC | |
1/16 in. |