Learn More
Ark-Plas Products™ PVC Tubing
Made of polyvinyl chloride. Ark-Plas Products™ Red PVC Hoses are ideal for use with connecting hoses of various diameters. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies.
Brand: Ark-Plas Products™ TCX29-FC1
Additional Details : Weight : 0.80000kg
Description
- Chemical and corrosion resistance
- Abrasion and wear resistance
- Rubber-like flexibility
Specifications
Black | |
PVC | |
3/8 in. |
1/4 in. | |
3/8 in. |