Learn More
Ark-Plas Products™ PVC Tubing
Made of polyvinyl chloride. Ark-Plas Products™ Red PVC Hoses are ideal for use with connecting hoses of various diameters. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies.
Brand:  Ark-Plas Products™ TCX29-FC1
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11797719
  • 2688.98 SEK / Each
Estimated Shipment
30-06-2021
Add to basket
Description
Description
  • Chemical and corrosion resistance
  • Abrasion and wear resistance
  • Rubber-like flexibility
Specifications
Specifications
Black
PVC
3/8 in.
1/4 in.
3/8 in.