Läs mer
PVRIG Antibody (2334B), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB93652MFV450
4840.75 SEK valid until 2025-12-16
BÄSTA PRIS på kampanj! Använd kampanjkod: "24090" för att få ditt kampanjpris.
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PVRIG Monoclonal antibody specifically detects PVRIG in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOF
Specifikationer
| 50mM Sodium Borate | |
| C7orf15MGC138295, MGC104322, MGC138297, MGC2463, poliovirus receptor related immunoglobulin domain containing, Poliovirus receptor-related immunoglobulin domain-containing protein, transmembrane protein PVRIG | |
| Primary | |
| Store at 4C in the dark. |