Läs mer
Pyrazinoic Acid Ethyl Ester, TRC
Produktkod. 30417677
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30417677 100 mg
1 options
Produktkod. 30417677 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP841000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyrazinoic Acid Ethyl Ester
CAS 6924-68-1
Molekylformel C7H8N2O2
Kvantitet 100 mg
Synonym Ethyl 2-Pyrazinecarboxylate, Ethyl Pyrazinecarboxylate, Ethyl Pyrazinoate
InChI formel InChI=1S/C7H8N2O2/c1-2-11-7(10)6-5-8-3-4-9-6/h3-5H,2H2,1H3
LEDER CCOC(=O)c1cnccn1
IUPAC-namn ethyl pyrazine-2-carboxylate
Molekylvikt (g/mol) 152.15
Formel vikt 152.0586