Läs mer
Pyrazole N-Demethyl Sildenafil, TRC
Produktkod. 30424474
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424474 10 mg
30412426 1 mg
30418611 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424474 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP84280010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139755-95-6
Molekylformel C21H28N6O4S
Molekylvikt (g/mol) 460.55
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-3-propyl-, Piperazine, 1-[[3-(4,7-dihydroxy-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methyl- (9CI), 1H-Pyrazolo[4,3-d]pyrimidine, piperazine deriv., 5-[2-Ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-Ethoxy-5-(4-methylpiperazin-1-ylsulfonyl)phenyl]-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Desmethylsildenafil
LEDER CCCc1n[nH]c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4
Specifikationer
Kemiskt namn eller material Pyrazole N-Demethyl Sildenafil
Kvantitet 10 mg
InChI formel InChI=1S/C21H28N6O4S/c1-4-6-16-18-19(25-24-16)21(28)23-20(22-18)15-13-14(7-8-17(15)31-5-2)32(29,30)27-11-9-26(3)10-12-27/h7-8,13H,4-6,9-12H2,1-3H3,(H,24,25)(H,22,23,28)
Formel vikt 460.1893