missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, is used as a chemicals, pharmaceutical intermediates, reagents. It is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, is used as a chemicals, pharmaceutical intermediates, reagents. It is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | ∽180°C (decomposition) |
Quantity | 250 mg |
Solubility Information | Soluble in water |
Formula Weight | 163.14 |
Percent Purity | 97% |
Chemical Name or Material | Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'