Läs mer
Pyrethrin 2 (~30%, Technical Grade, Contains Pyrethin 1), TRC
Produktkod. 18063630
Change view
Kvantitet:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18063630 10 g
17969629 1 g
18041330 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18063630 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99124010G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 121-29-9
Molekylformel C22 H28 O5
Molekylvikt (g/mol) 372.45
Synonym Cyclopropanecarboxylic acid, 3-[(1E)-3-methoxy-2-methyl-3-oxo-1-propen-1-yl]-2,2-dimethyl-, (1S)-2-methyl-4-oxo-3-(2Z)-2,4-pentadien-1-yl-2-cyclopenten-1-yl ester, (1R,3R)-, Cyclopropanecarboxylic acid, 3-[(1E)-3-methoxy-2-methyl-3-oxo-1-propenyl]-2,2-dimethyl-, (1S)-2-methyl-4-oxo-3-(2Z)-2,4-pentadienyl-2-cyclopenten-1-yl ester, (1R,3R)- (9CI), Pyrethrin II (6CI), (+)-Pyrethronyl (+)-pyrethrate, Biospray S, Pyrethrin, Pyrethrin 2, Pyrethrum mist
IUPAC-namn [(1S)-2-methyl-4-oxo-3-[(2Z)-penta-2,4-dienyl]cyclopent-2-en-1-yl] (1R,3R)-3-[(E)-3-methoxy-2-methyl-3-oxoprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate
LEDER COC(=O)\C(=C\[C@@H]1[C@@H](C(=O)O[C@H]2CC(=O)C(=C2C)C\C=C/C=C)C1(C)C)\C
Specifikationer
Kemiskt namn eller material Pyrethrin II
Kvantitet 10 g
InChI formel InChI=1S/C22H28O5/c1-7-8-9-10-15-14(3)18(12-17(15)23)27-21(25)19-16(22(19,4)5)11-13(2)20(24)26-6/h7-9,11,16,18-19H,1,10,12H2,2-6H3/b9-8-,13-11+/t16-,18+,19+/m1/s1
Formel vikt 372.1937