missing translation for 'onlineSavingsMsg'
Läs mer

Pyrethrum Extract (Technical Grade), TRC

Produktkod. 17981185
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
17981185 10 mg
17971485 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17981185 Supplier Toronto Research Chemicals Supplier No. TRCP99124810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8003-34-7
Molekylformel C43H56O8
Synonym Pyrethrins, Pyrethrins and Pyrethroids, Biopiren plus, Chrysanthemates pyrethrins and pyrethroids, Diatect, Firmotox, Goldgeist forte, Nettafid, Parexan, Piresan, Pyganic, Pyganic MUP 20, Pyre-sin Plus, Pyrellin, Pyrethrine, Pyrethrines, Pyrethrins and Pyrethroids, chrysanthemates, Pyrethrins and Pyrethroids, esters, Pyrethrins, chrysanthemates, Pyrethro Vioryl 5SC, Pyrethrum, Pyrethrum (insecticide), Pyrethrum FS, Pyrevert, Pyrocide 50, Spruzit, Take Down, Take Down Garden Spray
LEDER C/C(C)=C/C1C(C)(C)[C@@H]1C(O[C@@H]2C(C)=C(C/C=C/C=C)C(C2)=O)=O.C/C(C([CH3]=O)=O)=C\C3C(C)(C)[C@@H]3C(OC4C(C)=C(C/C=C/C=C)C(C4)=O)=O

Specifications

Kemiskt namn eller material Pyrethrins
Kvantitet 10 mg
InChI formel InChI=1S/C22H28O5.C21H28O3/c1-7-8-9-10-15-14(3)18(12-17(15)23)27-21(25)19-16(22(19,4)5)11-13(2)20(24)26-6;1-7-8-9-10-15-14(4)18(12-17(15)22)24-20(23)19-16(11-13(2)3)21(19,5)6/h7-9,11,16,18-19H,1,10,12H2,2-6H3;7-9,11,16,18-19H,1,10,12H2,2-6H3/b9-8+,13-11+;9-8+/t16?,18?,19-;16?,18-,19-/m00/s1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.