Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Useful where conventional filter papers would be destroyed by acids and alkalis
Brand:  Quickfit VB30/06M
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 15558000
  • 1843.00 SEK / Pack of 5
Estimated Shipment
28-12-2020
Add to basket
Description
Description
  • Büchner vacuum filtration funnel with glass sintered disc
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • Useful where conventional filter papers would be destroyed by acids and alkalis
  • With a Quickfit™ ground glass cone to allow direct filtration into Quickfit™ Büchner flasks
Specifications
Specifications
ISO 3585, DIN 12217, ASTM E-438
Quickfit™ ground Glass cone
30 mL
Borosilicate Glass