Learn More
Pyrex™ Scoop with Hollow Stem
Product Code. 12631992
Change view
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Height (Metric):
70 mm
85 mm
100 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Unit Size
12631992 3 mL 70 mm Pack of 10
12651992 10 mL 100 mm Pack of 10
11970885 6 mL 85 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12631992 Supplier Pyrex Supplier No. 2115/02M
  • ONLINE EXCLUSIVE PRICE
    2580.00 SEK
    2085.24 SEK / Paket med 10
    Save 494.76 SEK
    19% Av
Beräknad leverans: 04-03-2027
Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.
  • With hollow stem to aid introduction of the powder into narrow neck vessels e.g., volumetric flasks
  • The flat base ensures stability on the balance pan
Specifications
Capacity (Metric) 3 mL
Material Borosilicate Glass
Height (Metric) 70 mm
Product Type Weighing scoop