Läs mer
Pyribambenz-Isopropyl, TRC
Produktkod. 30421248
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421248 250 mg
30410283 500 mg
30410915 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421248 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP420130250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 420138-41-6
Molekylformel C23 H25 N3 O5
Molekylvikt (g/mol) 423.4617
Synonym Pyribambenz-isopropyl, Benzoic acid, 4-[[[2-[(4,6-dimethoxy-2-pyrimidinyl)oxy]phenyl]methyl]amino]-, 1-methylethyl ester, 4-[[[2-[(4,6-Dimethoxy-2-pyrimidinyl)oxy]phenyl]methyl]amino]benzoic acid 1-methylethyl ester, Pyribambenz-isopropyl, ZJ 0702
IUPAC-namn propan-2-yl 4-[[2-(4,6-dimethoxypyrimidin-2-yl)oxyphenyl]methylamino]benzoate
LEDER COc1cc(OC)nc(Oc2ccccc2CNc3ccc(cc3)C(=O)OC(C)C)n1
Specifikationer
Kemiskt namn eller material Pyribambenz-isopropyl
Kvantitet 250 mg
InChI formel InChI=1S/C23H25N3O5/c1-15(2)30-22(27)16-9-11-18(12-10-16)24-14-17-7-5-6-8-19(17)31-23-25-20(28-3)13-21(26-23)29-4/h5-13,15,24H,14H2,1-4H3
Formel vikt 423.1794