Läs mer
Pyribambenz-Propyl, TRC
Produktkod. 30412620
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412620 250 mg
30424895 50 mg
30422403 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30412620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP420135250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 420138-40-5
Molekylformel C23 H25 N3 O5
Molekylvikt (g/mol) 423.46
Synonym Pyribambenz-propyl, Benzoic acid, 4-[[[2-[(4,6-dimethoxy-2-pyrimidinyl)oxy]phenyl]methyl]amino]-, propyl ester, 4-[[[2-[(4,6-Dimethoxy-2-pyrimidinyl)oxy]phenyl]methyl]amino]benzoic acid propyl ester, Propyl 4-[2-(4,6-dimethoxypyrimidin-2-yloxy)benzylamino]benzoate, Pyribambenz-propyl, ZJ 0273
IUPAC-namn propyl 4-[[2-(4,6-dimethoxypyrimidin-2-yl)oxyphenyl]methylamino]benzoate
LEDER CCCOC(=O)c1ccc(NCc2ccccc2Oc3nc(OC)cc(OC)n3)cc1
Specifikationer
Kemiskt namn eller material Pyribambenz-propyl
Kvantitet 250 mg
InChI formel InChI=1S/C23H25N3O5/c1-4-13-30-22(27)16-9-11-18(12-10-16)24-15-17-7-5-6-8-19(17)31-23-25-20(28-2)14-21(26-23)29-3/h5-12,14,24H,4,13,15H2,1-3H3
Formel vikt 423.1794