missing translation for 'onlineSavingsMsg'
Learn More

Pyribambenz-Propyl, TRC

Product Code. 30424895
Change view
Click to view available options
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30424895 50 mg
30412620 250 mg
30422403 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424895 Supplier Toronto Research Chemicals Supplier No. TRCP42013550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420138-40-5
Molekylformel C23 H25 N3 O5
Molekylvikt (g/mol) 423.46
Synonym Pyribambenz-propyl, Benzoic acid, 4-[[[2-[(4,6-dimethoxy-2-pyrimidinyl)oxy]phenyl]methyl]amino]-, propyl ester, 4-[[[2-[(4,6-Dimethoxy-2-pyrimidinyl)oxy]phenyl]methyl]amino]benzoic acid propyl ester, Propyl 4-[2-(4,6-dimethoxypyrimidin-2-yloxy)benzylamino]benzoate, Pyribambenz-propyl, ZJ 0273
IUPAC-namn propyl 4-[[2-(4,6-dimethoxypyrimidin-2-yl)oxyphenyl]methylamino]benzoate
LEDER CCCOC(=O)c1ccc(NCc2ccccc2Oc3nc(OC)cc(OC)n3)cc1

Specifications

Kemiskt namn eller material Pyribambenz-propyl
Kvantitet 50 mg
InChI formel InChI=1S/C23H25N3O5/c1-4-13-30-22(27)16-9-11-18(12-10-16)24-15-17-7-5-6-8-19(17)31-23-25-20(28-2)14-21(26-23)29-3/h5-12,14,24H,4,13,15H2,1-3H3
Formel vikt 423.1794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.