Läs mer
Pyridin-3-yl Dimethylcarbamate, Mikromol™
Produktkod. 18002300
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
18002300 100 mg
1 options
Produktkod. 18002300 Leverantör Mikromol Leverantörsnummer MM0429.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pyridin-3-yl Dimethylcarbamate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyridin-3-yl Dimethylcarbamate
CAS 51581-32-9
Analyt- eller komponentnamn Pyridin-3-yl Dimethylcarbamate
Frakt skick Room Temperature
Molekylformel C8 H10 N2 O2
Certifikat och efterlevnad ISO 9001
Synonym Carbamic acid, N,N-dimethyl-, 3-pyridinyl ester, Carbamic acid, dimethyl-, 3-pyridinyl ester (9CI), Carbamic acid, dimethyl-, 3-pyridyl ester (6CI,7CI), 3-(Dimethylcarbamoyloxy)pyridine, NSC 168373, Norpyridostigmine, Ro 1-5142, Pyridostigmine Related Compound A, Pyridostigmine Bromide Impurity A, Pyridostigmine Impurity A
InChI formel InChI=1S/C8H10N2O2/c1-10(2)8(11)12-7-4-3-5-9-6-7/h3-6H,1-2H3
LEDER CN(C)C(=O)Oc1cccnc1
IUPAC-namn pyridin-3-yl N,N-dimethylcarbamate
Molekylvikt (g/mol) 166.18
Kvantitet 100 mg
Formel vikt 166.0742 g/mol