Läs mer
Pyridine-2-carbonyl Chloride Hydrochloride, TRC
Produktkod. 30410690
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30410690 10 g
1 options
Produktkod. 30410690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99161010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyridine-2-carbonyl Chloride Hydrochloride
CAS 39901-94-5
Molekylformel C6H5Cl2NO
Kvantitet 10 g
Synonym 2-Pyridinecarbonyl Chloride Hydrochloride, 2-Picolinoyl Chloride Hydrochloride, 2-Pyridinoyl Chloride Hydrochloride, Picolinic Acid Chloride Hydrochloride, Picolinoyl Chloride Hydrochloride
InChI formel InChI=1S/C6H4ClNO.ClH/c7-6(9)5-3-1-2-4-8-5;/h1-4H;1H
LEDER Cl.ClC(=O)c1ccccn1
IUPAC-namn pyridine-2-carbonyl chloride;hydrochloride
Molekylvikt (g/mol) 178.02
Formel vikt 176.9748