Läs mer
Pyridine-2-carboxaldehyde Methiodide, TRC
Produktkod. 30410144
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410144 100 mg
30414373 250 mg
30419483 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410144 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP991803100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3784-97-2
Molekylformel C7H8INO
Molekylvikt (g/mol) 249.05
Synonym 2-Formyl-1-methylpyridinium Iodide, PCAM
IUPAC-namn 1-methylpyridin-1-ium-2-carbaldehyde;iodide
LEDER O=CC1=[N+](C)C=CC=C1.[I-]
Specifikationer
Kemiskt namn eller material Pyridine-2-carboxaldehyde Methiodide
Kvantitet 100 mg
InChI formel InChI=1S/C7H8NO.HI/c1-8-5-3-2-4-7(8)6-9;/h2-6H,1H3;1H/q+1;/p-1
Formel vikt 248.9651