missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7080916
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
872-85-5
C6H5NO
107.112
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
872-85-5
C6H5NO
107.112
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O

Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

Solubility
Soluble in water (20 mg/ml at 20°C), ethanol and acetone.

Notes
It is air and light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -4°C
Density 1.132
Boiling Point 198°C
Flash Point 75°C (167°F)
Refractive Index 1.544
Quantity 25 g
Beilstein 105342
Sensitivity Air and light sensitive
Solubility Information Soluble in water (20mg/ml at 20°C),ethanol and acetone.
Formula Weight 107.11
Percent Purity 97%
Chemical Name or Material Pyridine-4-carboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'