Läs mer
Pyridine-iodine Chloride Complex, TRC
Produktkod. 30421229
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421229 50 mg
30425507 100 mg
30427747 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99892350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6443-90-9
Molekylformel C5H5N.ClI
Molekylvikt (g/mol) 241.46
LEDER ClI.c1ccncc1
Specifikationer
Kemiskt namn eller material Pyridine-iodine Chloride Complex
Kvantitet 50 mg
InChI formel InChI=1S/C5H5N.ClI/c1-2-4-6-5-3-1;1-2/h1-5H;
Formel vikt 240.9155