missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Catalog Number p-4408289
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
1049
CHEBI:16227
pyridine
C1=CC=NC=C1
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
1049
CHEBI:16227
pyridine
C1=CC=NC=C1

Pyridine is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 42°C
Color Colorless
Appearance Colorless liquid
Density 0.978
Boiling Point 115°C
Quantity 250 mL
Flash Point 20°C (68°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.51
Assay Percent Range >99.5%
Vapor Pressure 20 hPa (15mm Hg) at 20°C
Beilstein 103233
Solubility Information Miscible with water,alcohol,ether,pet ether,and oils.
Formula Weight 79.1
Grade Spectrophotometric
Chemical Name or Material Pyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'