Läs mer
Pyridinium Pentafluoroiodide - Hydrogenfluoride Complex, TRC
Produktkod. 30419655
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30419655 250 mg
1 options
Produktkod. 30419655 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP366480250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyridinium Pentafluoroiodide - Hydrogenfluoride Complex
CAS 2243786-10-7
Molekylformel C5H5N•(F6I)•H
Kvantitet 250 mg
Synonym Hexafluoroiodate(1-) Compound with Pyridine Conjugate Acid, IF5-pyridine-HF
InChI formel InChI=1S/C5H5N.F6I/c1-2-4-6-5-3-1;1-7(2,3,4,5)6/h1-5H;/q;-1/p+1
LEDER [F-][I+5]([F-])([F-])([F-])([F-])[F-].C1=CN=CC=C1.[H+]
Molekylvikt (g/mol) 321.01
Formel vikt 320.95