Läs mer
Pyridoxal 5-Phosphate Hydrate, TRC
Produktkod. 30413666
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30413666 250 mg
1 options
Produktkod. 30413666 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP992088250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyridoxal-5-phosphate hydrate
CAS 853645-22-4
Molekylformel C8 H10 N O6 P . x H2 O
Kvantitet 250 mg
Synonym 3-Hydroxy-2-methyl-5-[(phosphonooxy)methyl]-4]pyridinecarboxaldehyde hydrate
InChI formel InChI=1S/C8H10NO6P/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14/h2-3,11H,4H2,1H3,(H2,12,13,14)/i3D,4D2
LEDER [2H]C(=O)c1c(O)c(C)ncc1C([2H])([2H])OP(=O)(O)O
IUPAC-namn (4-formyl-5-hydroxy-6-methyl-3-pyridyl)methyl dihydrogen phosphate;hydrate
Molekylvikt (g/mol) 247.14
Formel vikt 250.0434