missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal Hydrochloride, Fisher BioReagents

Catalog Number p-28011
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
This item is not returnable. View return policy
65-22-5
C8H10ClNO3
203.622
FCHXJFJNDJXENQ-UHFFFAOYSA-N
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
6171
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
CC1=NC=C(C(=C1O)C=O)CO.Cl
This item is not returnable. View return policy
65-22-5
C8H10ClNO3
203.622
FCHXJFJNDJXENQ-UHFFFAOYSA-N
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
6171
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
CC1=NC=C(C(=C1O)C=O)CO.Cl
Pyridoxal Hydrochloride is useful for labeling amino acids and for their detection in the picomole range.
TRUSTED_SUSTAINABILITY
Melting Point 171°C
Color White
pH 2.1
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 8091
Formula Weight 203.63
Physical Form Powder or Solid
Chemical Name or Material Pyridoxal Hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'