Learn More
Pyridoxal Hydrochloride, TRC
Product Code. 30005803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005803 5 g 5g
30010216 25 g 25g
17961595 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005803 Supplier Toronto Research Chemicals Supplier No. TRCP9917205G
 Restricted Item
Beräknad leverans: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65-22-5
Molecular Formula C8 H9 N O3 . Cl H
Molecular Weight (g/mol) 203.62
Synonym 4-Pyridinecarboxaldehyde, 3-hydroxy-5-(hydroxymethyl)-2-methyl-, hydrochloride (9CI), Pyridoxal, hydrochloride (7CI,8CI), 2-Methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, 3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-Hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES Cl.Cc1ncc(CO)c(C=O)c1O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H
Formula Weight 203.0349
Chemical Name or Material Pyridoxal Hydrochloride