Läs mer
Pyridoxine EP Impurity A, Mikromol™
Produktkod. 17992496
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17992496 100 mg
17962056 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17992496 Leverantör Mikromol Leverantörsnummer MM0021.11
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pyridoxine EP Impurity A
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
CAS 5196-20-3
Analyt- eller komponentnamn 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
Frakt skick Room Temperature
Molekylformel C8 H9 N O2
Certifikat och efterlevnad ISO 17025
Synonym 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol, 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol, Pyridoxine Hydrochloride Imp. A (EP), 1,3-Dihydro-6-methylfuro[3,4-c]pyridin-7-ol, Pyridoxine Imp. A (EP), Pyridoxine Hydrochloride Impurity A, Pyridoxine Impurity A
InChI formel InChI=1S/C8H9NO2/c1-5-8(10)7-4-11-3-6(7)2-9-5/h2,10H,3-4H2,1H3
LEDER Cc1ncc2COCc2c1O
IUPAC-namn 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol
Molekylvikt (g/mol) 151.16
Kvantitet 100 mg
Formel vikt 151.0633 g/mol