missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine hydrochloride, 98%, MP Biomedicals™

Catalog Number p-4403658
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
58-56-0
C8H12ClNO3
205.638
MFCD00012807
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
6019
CHEBI:30961
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
CC1=NC=C(C(=C1O)CO)CO.Cl
This item is not returnable. View return policy
58-56-0
C8H12ClNO3
205.638
MFCD00012807
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
6019
CHEBI:30961
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
CC1=NC=C(C(=C1O)CO)CO.Cl
Color White
Density 1.43g/mL
Assay Percent Range >98%
Quantity 10 g
Formula Weight 205.64
Percent Purity 0.98
Physical Form Powder
Chemical Name or Material Pyridoxine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'