Läs mer
Pyridoxine, TRC
Produktkod. 17931705
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17931705 5 g
17902805 250 mg
17914925 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17931705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9917335G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65-23-6
Molekylformel C8 H11 N O3
Molekylvikt (g/mol) 169.18
Synonym 3,4-Pyridinedimethanol, 5-hydroxy-6-methyl-, Pyridoxol (8CI), 5-Hydroxy-6-methyl-3,4-pyridinedimethanol, 2-Methyl-3-hydroxy-4,5-di(hydroxymethyl)pyridine, 2-Methyl-4,5-bis(hydroxymethyl)-3-hydroxypyridine, Adermin, Bezatin, Pirivitol, Pyridoxin, Pyridoxine
IUPAC-namn 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol
LEDER Cc1ncc(CO)c(CO)c1O
Specifikationer
Kemiskt namn eller material Vitamin B6 (Pyridoxine)
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C8H11NO3/c1-5-8(12)7(4-11)6(3-10)2-9-5/h2,10-12H,3-4H2,1H3
Formel vikt 169.0739
Procent renhet >95%