Läs mer
Pyridoxol 5'-Phosphate >90%, TRC
Produktkod. 17912505
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912505 10 mg
17942605 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17912505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99178010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 447-05-2
Molekylformel C8 H12 N O6 P
Molekylvikt (g/mol) 249.16
Synonym 3,4-Pyridinedimethanol, 5-hydroxy-6-methyl-, α3-(dihydrogen phosphate) (9CI), Pyridoxol, 5-(dihydrogen phosphate) (6CI,7CI,8CI), NSC 83119, Pyridoxine 5-phosphate, Pyridoxine 5'-phosphate, Pyridoxine phosphate, Pyridoxol 5'-phosphate, [[5-Hydroxy-4-(hydroxymethyl)-6-methylpyridin-3-yl]methoxy]phosphonic acid, Pyridoxine 5'-Phosphate
IUPAC-namn [5-hydroxy-4-(hydroxymethyl)-6-methylpyridin-3-yl]methyl dihydrogen phosphate
LEDER Cc1ncc(COP(=O)(O)O)c(CO)c1O
Specifikationer
Kemiskt namn eller material Pyridoxol 5-Phosphate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C8H12NO6P/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14/h2,10-11H,3-4H2,1H3,(H2,12,13,14)
Formel vikt 249.0402
Procent renhet >95%