missing translation for 'onlineSavingsMsg'
Learn More

Pyriftalid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30322793
Change view
Click to view available options
Kvantitet:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30322793 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322793 Supplier Dr. Ehrenstorfer Supplier No. DREA16656000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Pyriftalid
CAS 135186-78-6
Analyt- eller komponentnamn Pyriftalid
Frakt skick Room Temperature
Molekylformel C15 H14 N2 O4 S
Koncentration 100 μg/mL
Certifikat och efterlevnad ISO 17034
InChI formel InChI=1S/C15H14N2O4S/c1-8-9-5-4-6-10(13(9)14(18)21-8)22-15-16-11(19-2)7-12(17-15)20-3/h4-8H,1-3H3
LEDER COc1cc(OC)nc(Sc2cccc3C(C)OC(=O)c23)n1
IUPAC-namn 7-(4,6-dimethoxypyrimidin-2-yl)sulfanyl-3-methyl-3H-2-benzofuran-1-one
Lösningsmedel eller matris Acetonitrile
Molekylvikt (g/mol) 318.35
Kvantitet 1 ml
Formel vikt 318.0674 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.