Läs mer
Pyrilamine Maleate Salt, TRC
Produktkod. 30179801
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179801 1 g
30180029 100 mg
30180389 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179801 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9972251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59-33-6
Molekylformel C17 H23 N3 O . C4 H4 O4
Molekylvikt (g/mol) 401.46
Synonym 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (2Z)-2-butenedioate (1:1) (9CI), 1,2-Ethanediamine, N-[(4-methoxyphenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, (Z)-2-butenedioate (1:1), Pyridine, 2-[[2-(dimethylamino)ethyl](p-methoxybenzyl)amino]-, maleate (1:1) (8CI), 2-[(2-Dimethylaminoethyl)(p-methoxybenzyl)amino]-pyridine maleate, 2-[[2-(Dimethylamino)ethyl](p-methoxybenzyl)amino]pyridine Bimaleate, AH, Anisopyradamine, Antamine, Anthisan maleate, Antihist, Antisan, Diaminide maleate, Enrumay, Histalet Forte, Histalon, Histan, Histapyran, Histatex, Histavet P, Mepyramine hydrogen maleinate, Mepyramine maleate, Minihist, N-p-Methoxybenzyl-N',N'-dimethyl-N-α-pyridylethylenediamine maleate, NSC 3604, Neo-Antergan maleate, Neoantergan maleate, Paramal, Paraminyl, Paraminyl maleate, Parmal, Prefrin A, Pymafed, Pyra Maleate, Pyramal, Pyranilamine maleate, Pyraninyl, Pyranisamine maleate, Pyrilamine maleate, Renstamin, Stamine, Stangen, Stangen maleate, Statomin maleate, Thylogen, Thylogen maleate
IUPAC-namn (Z)-but-2-enedioic acid;N'-[(4-methoxyphenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine
LEDER COc1ccc(CN(CCN(C)C)c2ccccn2)cc1.OC(=O)\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material Mepyramine maleate
Kvantitet 1 g
InChI formel InChI=1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vikt 401.1951