missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrimethamine, 100.5%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Color | White |
Assay Percent Range | 1.005 |
Quantity | 1 g |
Formula Weight | 248.7 |
Physical Form | Powder |
Chemical Name or Material | Pyrimethamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'