Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pyrimethanil |
| Förpackning | Glass Bottle |
| CAS | 53112-28-0 |
| Smältpunkt | 96.3°C |
| Synonym | 2-Anilino-4,6-dimethylpyrimidine |
| Industry Type | Environmental Analysis |
| Kvantitet | 100 mg |
| Kvalitet | Analytical Standard |